MAT047026653 Matrix Scientific

3-(4-Methoxy-phenyl)-2-phenyl-propylamine

  • MDL number MFCD03274501
  • Cas Number : 5636-50-0
  • Molecular Weight : 241.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42
Country of Origin USA
Boiling point 168-172�Torr
Product Link http://www.matrixscientific.com/008341.html

Safety Information

MSDS http://www.matrixscientific.com/008341.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart