MAT047026656 Matrix Scientific

2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)acetic acid

  • MDL number MFCD00664645
  • Cas Number : 39221-42-6
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-211
Country of Origin USA
Product Link http://www.matrixscientific.com/008344.html

Safety Information

MSDS http://www.matrixscientific.com/008344.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart