MAT047026671 Matrix Scientific

2-Isobutyl-quinoline-4-carboxylic acid

  • MDL number MFCD01548753
  • Cas Number : 24260-31-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192
Country of Origin USA
Product Link http://www.matrixscientific.com/008358.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008358.html
Availability details for
Quantity
  • Availability
  • Add to Cart