MAT047026672 Matrix Scientific

(2,5-Dioxo-1-phenyl-imidazolidin-4-yl)-acetic acid

  • MDL number MFCD01721303
  • Cas Number : 62848-47-9
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-229
Country of Origin USA
Product Link http://www.matrixscientific.com/008359.html

Safety Information

MSDS http://www.matrixscientific.com/008359.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart