MAT047026673 Matrix Scientific

3-(2-Phenyl-imidazol-1-yl)-propionic acid

  • MDL number MFCD03043651
  • Cas Number : 53660-14-3
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008360.html

Safety Information

MSDS http://www.matrixscientific.com/008360.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart