MAT047026685 Matrix Scientific

1-Cyano-4-fluoronaphthalene

  • MDL number MFCD00236656
  • Cas Number : 13916-99-9
  • Molecular Weight : 171.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/001742.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/001742.html
Availability details for
Quantity
  • Availability
  • Add to Cart