MAT047026691 Matrix Scientific

3-(2,3-Dihydro-benzo[1,4]dioxin-6-ylamino)-propionic acid

  • MDL number MFCD03701698
  • Cas Number : 436087-14-8
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008376.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008376.html
Availability details for
Quantity
  • Availability
  • Add to Cart