MAT047026707 Matrix Scientific

5-{[(2,5-Dimethyl-furan-3-carbonyl)-amino]-methyl}-furan-2-carboxylic acid

  • MDL number MFCD02737938
  • Cas Number : 385378-11-0
  • Molecular Weight : 263.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008390.html

Safety Information

MSDS http://www.matrixscientific.com/008390.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart