MAT047026715 Matrix Scientific

1-Naphthalen-1-yl-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD02197525
  • Cas Number : 340319-91-7
  • Molecular Weight : 255.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211
Country of Origin USA
Product Link http://www.matrixscientific.com/008397.html

Safety Information

MSDS http://www.matrixscientific.com/008397.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart