MAT047026730 Matrix Scientific

4-Amino-2-fluorophenol

  • MDL number MFCD00671760
  • Cas Number : 399-96-2
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/001746.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/001746.html
Availability details for
Quantity
  • Availability
  • Add to Cart