MAT047026736 Matrix Scientific

4-(5-Formyl-furan-2-yl)-2-hydroxy-benzoic acid

  • MDL number MFCD02738007
  • Cas Number : 436088-45-8
  • Molecular Weight : 232.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230�ec)
Country of Origin USA
Product Link http://www.matrixscientific.com/008419.html

Safety Information

MSDS http://www.matrixscientific.com/008419.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart