MAT047026762 Matrix Scientific

2-Imidazol-1-yl-phenylamine

  • MDL number MFCD03274531
  • Cas Number : 26286-54-4
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-104
Country of Origin USA
Product Link http://www.matrixscientific.com/008443.html

Safety Information

MSDS http://www.matrixscientific.com/008443.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart