MAT047026770 Matrix Scientific

3-(5-Thiophen-2-yl-1H-pyrrol-2-yl)-propionic acid

  • MDL number MFCD03269406
  • Cas Number : 432527-62-3
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008450.html

Safety Information

MSDS http://www.matrixscientific.com/008450.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart