MAT047026783 Matrix Scientific

5-Oxo-5-pyrrolidin-1-yl-pentanoic acid

  • MDL number MFCD01339572
  • Cas Number : 5724-80-1
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008462.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008462.html
Availability details for
Quantity
  • Availability
  • Add to Cart