MAT047026786 Matrix Scientific

1-Methylamino-3-phenoxy-propan-2-ol

  • MDL number MFCD01442095
  • Cas Number : 39631-73-7
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/008464.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008464.html
Availability details for
Quantity
  • Availability
  • Add to Cart