MAT047026795 Matrix Scientific

2-Chloro-N-[4-(1-pyrrolidinyl)phenyl]acetamide

  • MDL number MFCD00173016
  • Cas Number : 251097-15-1
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008473.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008473.html
Availability details for
Quantity
  • Availability
  • Add to Cart