MAT047026818 Matrix Scientific

1-(5-Methyl-benzo[1,2,5]thiadiazole-4-sulfonyl)-pyrrolidine-2-carboxylic acid

  • MDL number MFCD03044349
  • Cas Number : 436811-15-3
  • Molecular Weight : 327.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008492.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008492.html
Availability details for
Quantity
  • Availability
  • Add to Cart