MAT047026826 Matrix Scientific

[5-(4-Amino-phenyl)-4-methyl-4H-[1,2,4]triazol-3-ylsulfanyl]-acetic acid

  • MDL number MFCD02627932
  • Cas Number : 421567-49-9
  • Molecular Weight : 264.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008499.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008499.html
Availability details for
Quantity
  • Availability
  • Add to Cart