MAT047026834 Matrix Scientific

3-(2,6-Dimethyl-piperidin-1-yl)-propylamine

  • MDL number MFCD03445119
  • Cas Number : 85723-72-4
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 122-130�-32 Torr
Product Link http://www.matrixscientific.com/008506.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008506.html
Availability details for
Quantity
  • Availability
  • Add to Cart