MAT047026866 Matrix Scientific

5-Pyrrolidin-1-ylmethyl-furan-2-carboxylic acid

  • MDL number MFCD01821233
  • Cas Number : 400750-49-4
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008535.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008535.html
Availability details for
Quantity
  • Availability
  • Add to Cart