MAT047026903 Matrix Scientific

Furan-2-ylmethyl-(3-methyl-1-p-tolyl-but-3-enyl)-amine

  • MDL number MFCD03446187
  • Cas Number : 436087-19-3
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008569.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008569.html
Availability details for
Quantity
  • Availability
  • Add to Cart