MAT047026907 Matrix Scientific

(1-Furan-2-yl-2-phenyl-ethyl)-(4-methoxy-phenyl)-amine

  • MDL number MFCD03446188
  • Cas Number : 436087-20-6
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008573.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008573.html
Availability details for
Quantity
  • Availability
  • Add to Cart