MAT047026921 Matrix Scientific

(2-Isobutyrylamino-acetylamino)-acetic acid

  • MDL number MFCD03130024
  • Cas Number : 436096-89-8
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008584.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008584.html
Availability details for
Quantity
  • Availability
  • Add to Cart