MAT047026922 Matrix Scientific

N-(2-Azepan-1-yl-phenyl)-2-chloro-acetamide

  • MDL number MFCD03446301
  • Cas Number : 436087-22-8
  • Molecular Weight : 266.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/008585.html

Safety Information

MSDS http://www.matrixscientific.com/008585.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart