MAT047026931 Matrix Scientific

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene

  • MDL number MFCD00042090
  • Cas Number : 802-93-7
  • Molecular Weight : 410.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 99-100�mm
Product Link http://www.matrixscientific.com/001769.html

Safety Information

hcodes Corrosive/Irritant
MSDS http://www.matrixscientific.com/001769.html
Availability details for
Quantity
  • Availability
  • Add to Cart