MAT047026946 Matrix Scientific

(3-Methyl-2-oxo-2,3-dihydro-imidazo[1,2-a]pyridin-3-yl)-acetic acid

  • MDL number MFCD03274736
  • Cas Number : 436811-33-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008607.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008607.html
Availability details for
Quantity
  • Availability
  • Add to Cart