MAT047026947 Matrix Scientific

3-(4-Methoxy-phenyl)-2-phenyl-propionic acid

  • MDL number MFCD00046532
  • Cas Number : 4314-68-5
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Country of Origin USA
Product Link http://www.matrixscientific.com/008608.html

Safety Information

MSDS http://www.matrixscientific.com/008608.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart