MAT047026948 Matrix Scientific

3-(4-Fluoro-phenyl)-2-phenyl-propionic acid

  • MDL number MFCD03446669
  • Cas Number : 436086-86-1
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008609.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008609.html
Availability details for
Quantity
  • Availability
  • Add to Cart