MAT047027015 Matrix Scientific

4,6-Dimethyl-2-oxo-3,4-dihydro-2H-pyrido-[1,2-a]pyrimidine-4-carboxylic acid

  • MDL number MFCD03449006
  • Cas Number : 956783-58-7
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008671.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008671.html
Availability details for
Quantity
  • Availability
  • Add to Cart