MAT047027024 Matrix Scientific

Sodium 2-fluorobenzoate

  • MDL number MFCD01661442
  • Cas Number : 490-97-1
  • Molecular Weight : 162.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008684.html
Availability details for
Quantity
  • Availability
  • Add to Cart