MAT047027026 Matrix Scientific

5-(Pyridin-2-ylcarbamoyl)-3H-imidazole-4-carboxylic acid

  • MDL number MFCD01366409
  • Cas Number : 436088-75-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008002.html

Safety Information

MSDS http://www.matrixscientific.com/008002.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart