MAT047027042 Matrix Scientific

1-Bromo-2-iodotetrafluoroethane

  • MDL number MFCD00042136
  • Cas Number : 421-70-5
  • Molecular Weight : 306.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.648
Country of Origin USA
Boiling point 78
Product Link http://www.matrixscientific.com/001780.html

Safety Information

hcodes Toxic/Light Sensitive
MSDS http://www.matrixscientific.com/001780.html
Availability details for
Quantity
  • Availability
  • Add to Cart