MAT047027047 Matrix Scientific

2-Bromo-3,4,6-trifluoroaniline

  • MDL number MFCD00012370
  • Cas Number : 1481-21-6
  • Molecular Weight : 226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Purity 99
Country of Origin USA
Product Link http://www.matrixscientific.com/008708.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008708.html
Availability details for
Quantity
  • Availability
  • Add to Cart