MAT047027071 Matrix Scientific

4-Amino-2-chlorobenzotrifluoride

  • MDL number MFCD00278479
  • Cas Number : 445-13-6
  • Molecular Weight : 195.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 112-115
Product Link http://www.matrixscientific.com/008733.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008733.html
Availability details for
Quantity
  • Availability
  • Add to Cart