MAT047027087 Matrix Scientific

3-(Trifluoromethyl)benzoyl fluoride

  • MDL number MFCD00018047
  • Cas Number : 328-99-4
  • Molecular Weight : 192.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 159-163
Product Link http://www.matrixscientific.com/008748.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008748.html
Availability details for
Quantity
  • Availability
  • Add to Cart