MAT047027090 Matrix Scientific

3-(2-Amino-thiazol-4-yl)-chromen-2-one

  • MDL number MFCD00481614
  • Cas Number : 61636-28-0
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-227
Country of Origin US
Product Link http://www.matrixscientific.com/008009.html

Safety Information

MSDS http://www.matrixscientific.com/008009.html
Availability details for
Quantity
  • Availability
  • Add to Cart