MAT047027092 Matrix Scientific

3-Amino-3-(4-methoxy-1-naphthyl)-propanoic acid

  • MDL number MFCD02222925
  • Cas Number : 499987-13-2
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008010.html

Safety Information

MSDS http://www.matrixscientific.com/008010.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart