MAT047027104 Matrix Scientific

4-(Trifluoromethoxy)fluorobenzene

  • MDL number MFCD00040835
  • Cas Number : 352-67-0
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.323/20
Country of Origin USA
Boiling point 104-105
Product Link http://www.matrixscientific.com/008766.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/008766.html
Availability details for
Quantity
  • Availability
  • Add to Cart