MAT047027109 Matrix Scientific

3-(2-Thiophen-2-yl-acetylamino)-benzoic acid

  • MDL number MFCD01434377
  • Cas Number : 206983-05-3
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008016.html

Safety Information

MSDS http://www.matrixscientific.com/008016.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart