MAT047027120 Matrix Scientific

Methyl 2-methoxytetrafluoropropionate

  • MDL number MFCD00155951
  • Cas Number : 10186-63-7
  • Molecular Weight : 190.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.366/20
Country of Origin USA
Boiling point 40-41� Torr
Product Link http://www.matrixscientific.com/008786.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008786.html
Availability details for
Quantity
  • Availability
  • Add to Cart