MAT047027121 Matrix Scientific

2-Hydroxy-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD01529657
  • Cas Number : 328-90-5
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-178
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/008787.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008787.html
Availability details for
Quantity
  • Availability
  • Add to Cart