MAT047027128 Matrix Scientific

Allyl 2,2,3,3,3-pentafluoropropyl ether

  • MDL number MFCD02093320
  • Cas Number : 186907-75-5
  • Molecular Weight : 190.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 14
Country of Origin USA
Boiling point 85-86
Product Link http://www.matrixscientific.com/008793.html

Safety Information

hcodes Flammable/Irritant
MSDS http://www.matrixscientific.com/008793.html
Availability details for
Quantity
  • Availability
  • Add to Cart