MAT047027129 Matrix Scientific

Allyl 2,2,3,3-tetrafluoropropyl ether

  • MDL number MFCD02093319
  • Cas Number : 681-68-5
  • Molecular Weight : 172.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.183/25
Country of Origin USA
Boiling point 110�0mm
Product Link http://www.matrixscientific.com/008794.html

Safety Information

hcodes Flammable/Irritant
MSDS http://www.matrixscientific.com/008794.html
Availability details for
Quantity
  • Availability
  • Add to Cart