MAT047027147 Matrix Scientific

2,2-Bis(4-methylphenyl)hexafluoropropane

  • MDL number MFCD00042597
  • Cas Number : 1095-77-8
  • Molecular Weight : 332.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 98
Country of Origin USA
Boiling point 117�m
Product Link http://www.matrixscientific.com/008811.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008811.html
Availability details for
Quantity
  • Availability
  • Add to Cart