MAT047027160 Matrix Scientific

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside

  • MDL number MFCD00005666
  • Cas Number : 7240-90-6
  • Molecular Weight : 408.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-238
Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/008823.html

Safety Information

MSDS http://www.matrixscientific.com/008823.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart