MAT047027162 Matrix Scientific

1-Bromo-4-isobutylbenzene

  • MDL number MFCD03425886
  • Cas Number : 2051-99-2
  • Molecular Weight : 213.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -49
Purity 98
Density 1.2609/20
Country of Origin USA
Boiling point 97-98�Torr
Product Link http://www.matrixscientific.com/008824.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008824.html
Availability details for
Quantity
  • Availability
  • Add to Cart