MAT047027164 Matrix Scientific

6-Bromo-4-chloro-2-(trifluoromethyl)-quinoline

  • MDL number MFCD00153073
  • Cas Number : 1701-28-6
  • Molecular Weight : 310.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/008826.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008826.html
Availability details for
Quantity
  • Availability
  • Add to Cart