MAT047027177 Matrix Scientific

4-Bromo-2-fluorobenzylamine hydrochloride

  • MDL number MFCD00143266
  • Cas Number : 147181-08-6
  • Molecular Weight : 240.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254-257
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/008838.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008838.html
Availability details for
Quantity
  • Availability
  • Add to Cart