MAT047027181 Matrix Scientific

4-(Toluene-4-sulfonylamino)cyclohexane-carboxylic acid

  • MDL number MFCD01925898
  • Cas Number : 314042-69-8
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008026.html

Safety Information

MSDS http://www.matrixscientific.com/008026.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart