MAT047027193 Matrix Scientific

1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)-benzene

  • MDL number MFCD00042190
  • Cas Number : 68834-05-9
  • Molecular Weight : 273.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/008853.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008853.html
Availability details for
Quantity
  • Availability
  • Add to Cart